Chromoxane Cyanine R, pure
Chromoxane Cyanine R, pure
Chromoxane Cyanine R, pure
Chromoxane Cyanine R, pure
Thermo Scientific Chemicals
Chromoxane Cyanine R, pure
CAS: 3564-18-9 | C23H15Na3O9S
Have Questions?
Catalog NumberQuantity
ACR2151500505 g
Catalog number ACR215150050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialChromoxane Cyanine R
CAS3564-18-9
Molecular FormulaC23H15Na3O9S
MDL NumberMFCD00012411
Beilstein11, II, 240
View more
Indicator
RUO – Research Use Only