Chlorhexidine diacetate hydrate, 98%, Thermo Scientific Chemicals
Chlorhexidine diacetate hydrate, 98%, Thermo Scientific Chemicals
Chlorhexidine diacetate hydrate, 98%, Thermo Scientific Chemicals
Chlorhexidine diacetate hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chlorhexidine diacetate hydrate, 98%, Thermo Scientific Chemicals
CAS: 206986-79-0 | C26H38Cl2N10O4 | 625.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR214980250
Price (MYR)
1,112.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,112.00
Add to cart
Chemical Identifiers
CAS3251-69-2
IUPAC Name2-(1H-imidazol-5-yl)acetic acid hydrochloride
Molecular FormulaC5H7ClN2O2
InChI KeyMWHLCFYPFGFBQO-UHFFFAOYSA-N
SMILESCl.OC(=O)CC1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%