Molecular sieves 4A, powder <50 micron, Thermo Scientific Chemicals
Molecular sieves 4A, powder &lt;50 micron, Thermo Scientific Chemicals
Molecular sieves 4A, powder &lt;50 micron, Thermo Scientific Chemicals
Molecular sieves 4A, powder &lt;50 micron, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molecular sieves 4A, powder <50 micron, Thermo Scientific Chemicals
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Packaging:
Plastic Bottle
Catalog number ACR214800010
Price (MYR)
1,021.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,021.00
Add to cart
Chemical Identifiers
CAS96-33-3
IUPAC Namemethyl prop-2-enoate
Molecular FormulaC4H6O2
InChI KeyBAPJBEWLBFYGME-UHFFFAOYSA-N
SMILESCOC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.4010 to 1.4040 (20°C, 589 nm)
Color scale=<10 APHA
Infrared spectrumConforms
Water=<0.08 %
View more
Drying agent
RUO – Research Use Only