Molecular sieves 3A, powder <50 micron, Thermo Scientific Chemicals
Molecular sieves 3A, powder &lt;50 micron, Thermo Scientific Chemicals
Molecular sieves 3A, powder &lt;50 micron, Thermo Scientific Chemicals
Molecular sieves 3A, powder &lt;50 micron, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molecular sieves 3A, powder <50 micron, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number ACR214795000
Price (MYR)
806.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
806.00
Add to cart
Specifications
Chemical Name or MaterialMolecular sieves 3A
ColorUndesignated
PackagingGlass bottle
Particle Size500 Micron Max.
Residual Water< 1.5% (550°C, 2 hrs.)
View more
Drying agent
RUO – Research Use Only