3-Aminoquinoline, 99%, Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
CAS: 580-17-6 | C9H8N2 | 144.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR213780050
Price (MYR)
944.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
944.00
Add to cart
Chemical Identifiers
CAS132203-74-8
SpecificationsSpecification SheetSpecification Sheet
Formpowder
Water Content (Karl Fischer Titration)≤ 5.0%
Appearance (Color)white to cream or pale yellow
Assay (unspecified)> 97.5%
Optical Rotation-90? ? 3? (c=1, ethanol)