3-Aminoquinoline, 99%, Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
CAS: 580-17-6 | C9H8N2 | 144.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR213780010
Price (MYR)
457.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
457.00
Add to cart
Chemical Identifiers
CAS274-09-9
IUPAC Name2H-1,3-benzodioxole
Molecular FormulaC7H6O2
InChI KeyFTNJQNQLEGKTGD-UHFFFAOYSA-N
SMILESC1OC2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5370-1.5400 @ 20?C
Appearance (Color)Clear colorless to yellow to orange
Assay (GC)≥98.5%