Copper(I) bromide-dimethyl sulfide complex, 99%, Thermo Scientific Chemicals
Copper(I) bromide-dimethyl sulfide complex, 99%, Thermo Scientific Chemicals
Copper(I) bromide-dimethyl sulfide complex, 99%, Thermo Scientific Chemicals
Copper(I) bromide-dimethyl sulfide complex, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper(I) bromide-dimethyl sulfide complex, 99%, Thermo Scientific Chemicals
CAS: 54678-23-8 | C2H6BrCuS | 205.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR213180250
Price (MYR)
1,283.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,283.00
Add to cart
Chemical Identifiers
CAS7697-37-2
IUPAC Namenitric acid
Molecular FormulaHNO3
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILESO[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay from Supplier's CofA65.0-70.0%
Total Metal Impurities≤0.0001%