Naphthol AS, 99%
Naphthol AS, 99%
Naphthol AS, 99%
Thermo Scientific Chemicals
Naphthol AS, 99%
CAS: 92-77-3 | C17H13NO2 | 263.28 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR212181000100 gGlass bottle
Catalog number ACR212181000
Price (MYR)
617.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
617.00
Add to cart
Chemical Identifiers
CAS1192-30-9
IUPAC Name2-(bromomethyl)oxolane
Molecular FormulaC5H9BrO
InChI KeyVOHILFSOWRNVJJ-UHFFFAOYNA-N
SMILESBrCC1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
FormLiquid
Refractive Index1.4840-1.4900 @ 20?C
Appearance (Color)Clear colorless to yellow