Iron(III) citrate hydrate, 98%
Iron(III) citrate hydrate, 98%
Iron(III) citrate hydrate, 98%
Iron(III) citrate hydrate, 98%
Thermo Scientific Chemicals
Iron(III) citrate hydrate, 98%
CAS: 334024-15-6 | C6H5FeO7 | 244.94 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR2118500101 kgPlastic bottle
Catalog number ACR211850010
Price (MYR)
1,060.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,060.00
Add to cart
Chemical Identifiers
CAS16052-06-5
IUPAC Name3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
Molecular FormulaC9H20N2O4S
InChI KeyOWXMKDGYPWMGEB-UHFFFAOYSA-N
SMILESOCCN1CCN(CCCS(O)(=O)=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.5 %
UV(0.1 M in water) 1 cm cell
Appearance (Color)White
Appearance (Form)Crystalline powder
pH5 to 6 (10 % w/v at 20°C)