Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
CAS: 94-36-0 | C14H10O4 | 242.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR211781000
Price (MYR)
452.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
452.00
Add to cart
Chemical Identifiers
CAS96-30-0
IUPAC Name2-chloro-N-methylacetamide
Molecular FormulaC3H6ClNO
InChI KeyHOZLOOPIXHWKCI-UHFFFAOYSA-N
SMILESCNC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥96.0%
Melting Point (clear melt)38.0-47.0?C
Radical initiators
RUO – Research Use Only