Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
CAS: 94-36-0 | C14H10O4 | 242.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
100 g
Packaging:
Plastic bottle
Catalog number ACR211780010
Price (MYR)
1,305.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,305.00
Add to cart
Chemical Identifiers
CAS39234-83-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Melting Point (clear melt)62.0-68.0?C
FormPowder
Assay (GC)≥96.0%
Radical initiators
RUO – Research Use Only