Deuterium chloride, for NMR, 1M solution in D2O, 99.8 atom% D, AcroSeal™
Deuterium chloride, for NMR, 1M solution in D2O, 99.8 atom% D, AcroSeal™
Deuterium chloride, for NMR, 1M solution in D2O, 99.8 atom% D, AcroSeal™
Deuterium chloride, for NMR, 1M solution in D2O, 99.8 atom% D, AcroSeal™
Thermo Scientific Chemicals
Deuterium chloride, for NMR, 1M solution in D2O, 99.8 atom% D, AcroSeal™
CAS: 7698-05-7 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR210601000100 mLAcroSeal™ Glass Bottle
Catalog number ACR210601000
Price (MYR)
2,457.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
2,457.00
Add to cart
Chemical Identifiers
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<50 APHA
Water=<0.05 % (K.F.)
Infrared spectrumConforms
Specific gravity(20°C) 1.02 to 1.04
View more
For NMR
RUO – Research Use Only