Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals
Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals
Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals
Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals
Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals
CAS: 92112-69-1 | C6H14
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
Packaging:
Metal Drum
Catalog number ACR210590250
Price (MYR)
1,863.00
Add to cart
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (MYR)
1,863.00
Add to cart
Chemical Identifiers
CAS92112-69-1, 110-54-3
Specifications
Chemical Name or MaterialHexanes
Melting Point-95.0°C
Name NoteMixture of isomers
CAS92112-69-1
Health Hazard 1Danger