N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
10 L
25 L
Packaging:
View all
Glass Bottle
Catalog number ACR210580025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS504-78-9
IUPAC Name1,3-thiazolidine
Molecular FormulaC3H7NS
InChI KeyOGYGFUAIIOPWQD-UHFFFAOYSA-N
SMILESC1CSCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.5510-1.5560 @ 20?C
For analysis
RUO – Research Use Only