Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals
Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals
Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals
Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals
CAS: 593-56-6 | CH5NO·HCl | 83.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR210490250
Price (MYR)
1,288.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,288.00
Add to cart
Chemical Identifiers
CAS13726-76-6
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-5-[(diaminomethylidene)amino]pentanoic acid
Molecular FormulaC11H22N4O4
InChI KeyHSQIYOPBCOPMSS-ZETCQYMHSA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CCCN=C(N)N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental AnalysisC: 46.0 ? 1%, H: 8.0 ? 1%. N: 19.0 ? 1% Calc. for x 0.65 H₂O (4%)
Water Content (Karl Fischer Titration)≤5.0%
Optical Rotation|1²⁴ = -6 ? 2? (C=2 in AcOH)
Solution TestClear colorless solution (50 mg/mL in Water)
Assay from Supplier's CofAD-Enantiomer ≤ 0.1%
View more
Alkylation reagent
RUO – Research Use Only