(-)-Camphanic acid chloride, 98%, Thermo Scientific Chemicals
(-)-Camphanic acid chloride, 98%, Thermo Scientific Chemicals
(-)-Camphanic acid chloride, 98%, Thermo Scientific Chemicals
(-)-Camphanic acid chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(-)-Camphanic acid chloride, 98%, Thermo Scientific Chemicals
CAS: 39637-74-6 | C10H13ClO3 | 216.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number ACR209890250
Price (MYR)
5,462.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
5,462.00
Add to cart
Chemical Identifiers
CAS22282-69-5
IUPAC Name2-fluoro-1,4-dihydropyridin-4-one
Molecular FormulaC5H4FNO
InChI KeyBRLSDLHMGRDAPF-UHFFFAOYSA-N
SMILESFC1=CC(=O)C=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)143.0-152.0?C
Appearance (Color)White to pale cream
FormPowder
Chiral derivatization reagent
RUO – Research Use Only