Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
CAS: 1730-25-2 | C3H5BrMg | 145.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number ACR209531000
Price (MYR)
775.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
775.00
EA
Add to cart
Chemical Identifiers
CAS22934-41-4
IUPAC Namequinoline-5-carbaldehyde
Molecular FormulaC10H7NO
InChI KeyBNEVFKZLYCGDFG-UHFFFAOYSA-N
SMILESO=CC1=C2C=CC=NC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.0%
Free acid (titration)≤1.5%
Melting Point (clear melt)91.0-100.0?C