Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
CAS: 12501-23-4 | H5O41PW12 | 2880.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR208315000
Price (MYR)
3,758.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
3,758.00
Add to cart
Chemical Identifiers
CAS7778-50-9
IUPAC Namedipotassium [(oxidodioxochromio)oxy]chromiumoylolate
Molecular FormulaCr2K2O7
InChI KeyKMUONIBRACKNSN-UHFFFAOYSA-N
SMILES[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to orange-red
Insoluble matter=<0.05 % (in water)
Chloride (Cl)=<700 ppm
Appearance (Form)Crystals
Titration Iodometric>=99.4 %