Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
CAS: 12501-23-4 | H5O41PW12 | 2880.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR208310250
Price (MYR)
580.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
580.00
Add to cart
Chemical Identifiers
CAS298-07-7
IUPAC Namebis[(2-ethylhexyl)oxy]phosphinic acid
Molecular FormulaC16H35O4P
InChI KeySEGLCEQVOFDUPX-UHFFFAOYNA-N
SMILESCCCCC(CC)COP(O)(=O)OCC(CC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (Titration)≥94.0 to ≤106.0%
Impurity contentMonoethylhexylphosphoric acid: ≤3%
Refractive Index1.4400-1.4440 @ 20?C