Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
CAS: 12501-23-4 | H5O41PW12 | 2880.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR208310025
Price (MYR)
14,378.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
14,378.00
Add to cart
Specifications
Chemical Name or MaterialPhosphotungstic acid hydrate
Name Notep.a.
CAS12501-23-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement: