CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
Thermo Scientific Chemicals
CHES, 98+%, for biochemistry
CAS: 103-47-9 | C8H17NO3S | 207.288 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR208180250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS39959-54-1
IUPAC Name(3-bromophenyl)methanaminium
Molecular FormulaC7H9BrN
InChI KeySUYJXERPRICYRX-UHFFFAOYSA-O
SMILES[NH3+]CC1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤1.0%
Appearance (Color)White to cream
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
FormPowder or crystalline powder or crystals or flakes
Biological buffer
RUO – Research Use Only