Ascarite™ II, CO2 absorbent, 20-30 mesh, capacity: 40-50%, product of Arthur H. Thomas Company, Thermo Scientific Chemicals
Ascarite(II), CO2 absorbent, 20-30 mesh, capacity: 40-50%, CAS Number-81133-20-2, 1310-73-2, 7631-86-9, 100g, Beige to Brown, Danger, GHS H Statement: Causes severe skin burns and eye damage.
Have Questions?
Change view
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Catalog number ACR208091000
Price (MYR)
1,064.00
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,064.00
Add to cart
Chemical Identifiers
CAS81133-20-2, 1310-73-2, 7631-86-9
Specifications
Chemical Name or MaterialAscarite(II)™, CO2 absorbent, 20-30 mesh, capacity: 40-50%
ColorBeige to Brown
Physical FormGranular
Name Note20- to 30-mesh
CAS81133-20-2