Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Thermo Scientific Chemicals
Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Thermo Scientific Chemicals
CAS: 81133-20-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Catalog number ACR208085000
Price (MYR)
3,754.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
3,754.00
Add to cart
Chemical Identifiers
CAS63039-48-5
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}pent-4-ynoate
Molecular FormulaC10H14NO4
InChI KeyAMKHAJIFPHJYMH-ZETCQYMHSA-M
SMILESCC(C)(C)OC(=O)N[C@@H](CC#C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Specific optical rotation+24° to +28° (20°C, 589 nm) (C=1, Methanol)
Appearance (Color)White to light yellow
GC>=94 %
Gas Adsorbent
RUO – Research Use Only