Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Thermo Scientific Chemicals
Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Thermo Scientific Chemicals
CAS: 81133-20-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Catalog number ACR208080025
Price (MYR)
13,507.00
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (MYR)
13,507.00
Add to cart
Chemical Identifiers
CAS81133-20-2, 1310-73-2, 7631-86-9
Specifications
Chemical Name or MaterialAscarite(II)™, CO2 absorbent, 8 to 20 mesh
ColorBrown
Physical FormGranular
Name Note8- to 20-mesh
CAS81133-20-2
View more
Gas Adsorbent
RUO – Research Use Only