Barium iodide hydrate, 98%, for analysis
Barium iodide hydrate, 98%, for analysis
Barium iodide hydrate, 98%, for analysis
Barium iodide hydrate, 98%, for analysis
Barium iodide hydrate, 98%, for analysis
Barium iodide hydrate, 98%, for analysis
Barium iodide hydrate, 98%, for analysis
Thermo Scientific Chemicals
Barium iodide hydrate, 98%, for analysis
CAS: 85017-89-6 | BaI2 | 391.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR20800050050 gGlass bottle
Catalog number ACR208000500
Price (MYR)
933.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
933.00
Add to cart
Chemical Identifiers
CAS137-26-8
IUPAC NameN,N-dimethyl[(dimethylcarbamothioyl)disulfanyl]carbothioamide
Molecular FormulaC6H12N2S4
InChI KeyKUAZQDVKQLNFPE-UHFFFAOYSA-N
SMILESCN(C)C(=S)SSC(=S)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to cream
Assay (HPLC)≥96.0%