Sodium oxalate, 98.5%, extra pure, Thermo Scientific Chemicals
Sodium oxalate, 98.5%, extra pure, Thermo Scientific Chemicals
Sodium oxalate, 98.5%, extra pure, Thermo Scientific Chemicals
Sodium oxalate, 98.5%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium oxalate, 98.5%, extra pure, Thermo Scientific Chemicals
CAS: 62-76-0 | C2Na2O4 | 134.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR207725000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium oxalate
Name Notepure
CAS62-76-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: