Remazol Brilliant Blue R, pure
Remazol Brilliant Blue R, pure
Remazol Brilliant Blue R, pure
Remazol Brilliant Blue R, pure
Thermo Scientific Chemicals
Remazol Brilliant Blue R, pure
CAS: 2580-78-1 | C22H16N2Na2O11S3 | 626.53 g/mol
Have Questions?
Catalog NumberQuantity
ACR20755025025 g
Catalog number ACR207550250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialRemazol Brilliant Blue R
CAS2580-78-1
Molecular FormulaC22H16N2Na2O11S3
MDL NumberMFCD00001215
SynonymC.I. 61200