Spironolactone, 99%
Spironolactone, 99%
Spironolactone, 99%
Thermo Scientific Chemicals
Spironolactone, 99%
CAS: 52-01-7 | C24H32O4S | 416.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR207460050
Price (MYR)
1,648.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,648.00
Add to cart
Chemical Identifiers
CAS104-47-2
IUPAC Name2-(4-methoxyphenyl)acetonitrile
Molecular FormulaC9H9NO
InChI KeyPACGLQCRGWFBJH-UHFFFAOYSA-N
SMILESCOC1=CC=C(CC#N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5295-1.5335 @ 20?C
Appearance (Color)Clear colorless to pale yellow to yellow to brown