Procainamide hydrochloride, 99%, Thermo Scientific Chemicals
Procainamide hydrochloride, 99%, Thermo Scientific Chemicals
Procainamide hydrochloride, 99%, Thermo Scientific Chemicals
Procainamide hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Procainamide hydrochloride, 99%, Thermo Scientific Chemicals
CAS: 614-39-1 | C13H21N3O·HCl | 271.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR207300250
Price (MYR)
762.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
762.00
Add to cart
Chemical Identifiers
CAS7790-94-5
IUPAC Namesulfurochloridic acid
Molecular FormulaClHO3S
InChI KeyXTHPWXDJESJLNJ-UHFFFAOYSA-N
SMILESOS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Slightly hazy viscous liquid
Titration>=96.0 %