1-Phenyl-2-thiourea, 97%, Thermo Scientific Chemicals
1-Phenyl-2-thiourea, 97%, Thermo Scientific Chemicals
1-Phenyl-2-thiourea, 97%, Thermo Scientific Chemicals
1-Phenyl-2-thiourea, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Phenyl-2-thiourea, 97%, Thermo Scientific Chemicals
CAS: 103-85-5 | C7H8N2S | 152.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR207250050
Price (MYR)
425.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
425.00
Add to cart
Chemical Identifiers
CAS821-55-6
IUPAC Namenonan-2-one
Molecular FormulaC9H18O
InChI KeyVKCYHJWLYTUGCC-UHFFFAOYSA-N
SMILESCCCCCCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Refractive Index1.4195-1.4235 @ 20?C
Appearance (Color)Clear colorless