L(-)-Phenylephrine hydrochloride, 99%, Thermo Scientific Chemicals
L(-)-Phenylephrine hydrochloride, 99%, Thermo Scientific Chemicals
L(-)-Phenylephrine hydrochloride, 99%, Thermo Scientific Chemicals
L(-)-Phenylephrine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(-)-Phenylephrine hydrochloride, 99%, Thermo Scientific Chemicals
CAS: 61-76-7 | C9H14ClNO2 | 203.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number ACR207240100
Price (MYR)
744.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
744.00
Add to cart
Chemical Identifiers
CAS304-55-2
IUPAC Name2,3-disulfanylbutanedioic acid
Molecular FormulaC4H6O4S2
InChI KeyACTRVOBWPAIOHC-UHFFFAOYNA-N
SMILESOC(=O)C(S)C(S)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%