Murexide, ACS reagent
Murexide, ACS reagent
Murexide, ACS reagent
Murexide, ACS reagent
Thermo Scientific Chemicals
Murexide, ACS reagent
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
500 g
Packaging:
Glass Bottle
Catalog number ACR206890250
Price (MYR)
1,202.00
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,202.00
Add to cart
Chemical Identifiers
CAS99-81-0
IUPAC Name2-bromo-1-(4-nitrophenyl)ethan-1-one
Molecular FormulaC8H6BrNO3
InChI KeyMBUPVGIGAMCMBT-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C=C1)C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=94 %
Appearance (Color)Yellow to orange
Melting point94°C to 100°C
Indicator
RUO – Research Use Only