Murexide, ACS reagent
Murexide, ACS reagent
Murexide, ACS reagent
Murexide, ACS reagent
Thermo Scientific Chemicals
Murexide, ACS reagent
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Catalog number ACR206890050
Price (MYR)
520.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
520.00
Add to cart
Chemical Identifiers
CAS20596-34-3
IUPAC Nameplatinum(4+) dihydrogen hexabromide
Molecular FormulaBr6H2Pt
InChI KeyVOEHEFXBCNCSCK-UHFFFAOYSA-J
SMILES[H+].[H+].[Br-].[Br-].[Br-].[Br-].[Br-].[Br-].[Pt+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Indicator
RUO – Research Use Only