Iodic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Iodic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Iodic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Iodic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodic acid, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 7782-68-5 | ILiO3 | 181.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number ACR206450500
Price (MYR)
723.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
723.00
Add to cart
Specifications
Chemical Name or MaterialIodic acid
ColorWhite to Beige
Name Notep.a.
CAS7782-68-5
Melting Point110.0°C
View more
Oxidation reagent for analysis
RUO – Research Use Only