Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR206385000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPhosphomolybdic acid hydrate
ColorYellow
Name Noteca. 78% Phosphomolybdic Acid
CAS51429-74-4
Health Hazard 1GHS Signal Word: Danger
View more
Oxidation reagent
RUO – Research Use Only