Check Order Status
Ammonium nitrate, 99+%, for analysis
Element Search
Structure Search
Thermo Scientific Chemicals
Ammonium nitrate, 99+%, for analysis
Have Questions?
Catalog number
ACR205860025
Price (MYR)
1,431.00
Add to cart
Request bulk or custom format
Price (MYR)
1,431.00
Add to cart
Specifications
Melting Point
169°C
Boiling Point
210°C
Packaging
Plastic Bottle
Quantity
2.5 kg
Unit Size
Ammonium nitrate, 99+%, for analysis 2.5kg Plastic Bottle Cas No. 6484-52-2
RUO – Research Use Only