Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water, Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water, Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water, Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water, Thermo Scientific Chemicals
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass bottle
Catalog number ACR205840025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium hydroxide
Boiling Point38.0°C
CAS1336-21-6
Health Hazard 1Danger
Melting Point-57.0°C