2-Thiophenemethylamine, 95%, Thermo Scientific Chemicals
2-Thiophenemethylamine, 95%, Thermo Scientific Chemicals
2-Thiophenemethylamine, 95%, Thermo Scientific Chemicals
2-Thiophenemethylamine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Thiophenemethylamine, 95%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR205570250
Price (MYR)
828.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
828.00
Add to cart
Chemical Identifiers
CAS38183-03-8
IUPAC Name7,8-dihydroxy-2-phenyl-4H-chromen-4-one
Molecular FormulaC15H10O4
InChI KeyCOCYGNDCWFKTMF-UHFFFAOYSA-N
SMILESOC1=CC=C2C(=O)C=C(OC2=C1O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Pale yellow to yellow
Water Content (Karl Fischer Titration)3.42-9.61% (0.5-1.5 waters)
Assay (HPLC)≥96.0%
Melting Point (clear melt)242.0-256.0?C