Florisil™, 100-200 mesh
Florisil™, 100-200 mesh
Florisil™, 100-200 mesh
Florisil™, 100-200 mesh
Thermo Scientific Chemicals
Florisil™, 100-200 mesh
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
5 kg
Catalog number ACR205445000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS97-00-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or mass and/or
Appearance (Form)chunks
Infrared spectrumConforms
GC>=98.5 %
Melting point48°C to 53°C