D(+)-Glucuronic acid, sodium salt, monohydrate, 99%, Thermo Scientific Chemicals
D(+)-Glucuronic acid, sodium salt, monohydrate, 99%, Thermo Scientific Chemicals
D(+)-Glucuronic acid, sodium salt, monohydrate, 99%, Thermo Scientific Chemicals
D(+)-Glucuronic acid, sodium salt, monohydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
D(+)-Glucuronic acid, sodium salt, monohydrate, 99%, Thermo Scientific Chemicals
CAS: 14984-34-0 | C6H9NaO7·H2O | 234.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number ACR204570250
Price (MYR)
718.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
718.00
Add to cart
Specifications
Chemical Name or MaterialD(+)-Glucuronic acid, sodium salt, monohydrate
CAS14984-34-0
Melting Point134°C to 140°C
ColorWhite
EINECS Number239-065-7