Tetraphenylphosphonium bromide, 99%, Thermo Scientific Chemicals
Tetraphenylphosphonium bromide, 99%, Thermo Scientific Chemicals
Tetraphenylphosphonium bromide, 99%, Thermo Scientific Chemicals
Tetraphenylphosphonium bromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetraphenylphosphonium bromide, 99%, Thermo Scientific Chemicals
CAS: 2751-90-8 | C24H20BrP | 419.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number ACR204020250
Price (MYR)
593.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
593.00
Add to cart
Chemical Identifiers
CAS54451-25-1
IUPAC Namedicerium(3+) tricarbonate
Molecular FormulaC3Ce2O9
InChI KeyGHLITDDQOMIBFS-UHFFFAOYSA-H
SMILES[Ce+3].[Ce+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.001% max.
for C-C Bond Formation (Olefination)
RUO – Research Use Only