Cyclophosphamide monohydrate, 97%, Thermo Scientific Chemicals
Cyclophosphamide monohydrate, 97%, Thermo Scientific Chemicals
Cyclophosphamide monohydrate, 97%, Thermo Scientific Chemicals
Cyclophosphamide monohydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclophosphamide monohydrate, 97%, Thermo Scientific Chemicals
CAS: 6055-19-2 | C7H17Cl2N2O3P | 279.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR203960010
Price (MYR)
545.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
545.00
Add to cart
Specifications
Chemical Name or MaterialCyclophosphamide monohydrate
CAS6055-19-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause genetic defects.
May cause cancer.
May damage the unborn child.
Toxic if swallowed.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Wash face,hands and any exposed skin thoroughly after