Trimethylsilylacetylene, 98%, Thermo Scientific Chemicals
Trimethylsilylacetylene, 98%, Thermo Scientific Chemicals
Trimethylsilylacetylene, 98%, Thermo Scientific Chemicals
Trimethylsilylacetylene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trimethylsilylacetylene, 98%, Thermo Scientific Chemicals
CAS: 1066-54-2 | C5H10Si | 98.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR203570250
Price (MYR)
1,480.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,480.00
EA
Add to cart
Chemical Identifiers
CAS188111-79-7
IUPAC Nametert-butyl 3-aminopiperidine-1-carboxylate
Molecular FormulaC10H20N2O2
InChI KeyAKQXKEBCONUWCL-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCCC(N)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4710-1.4750 @ 20?C
Optical Rotation-27.5 ± 1.5? (neat)