Trimethylsilylacetylene, 98%, Thermo Scientific Chemicals
Trimethylsilylacetylene, 98%, Thermo Scientific Chemicals
Trimethylsilylacetylene, 98%, Thermo Scientific Chemicals
Trimethylsilylacetylene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trimethylsilylacetylene, 98%, Thermo Scientific Chemicals
CAS: 1066-54-2 | C5H10Si | 98.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR203570050
Price (MYR)
601.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
601.00
Add to cart
Chemical Identifiers
CAS75-54-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (unspecified)≥97.0%