Vincristine sulfate, 95%
Vincristine sulfate, 95%
Vincristine sulfate, 95%
Vincristine sulfate, 95%
Thermo Scientific Chemicals
Vincristine sulfate, 95%
CAS: 2068-78-2 | C46H56N4O10·H2SO4 | 923.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR2034400505 mgGlass Bottle
Catalog number ACR203440050
Price (MYR)
1,178.00
EA
Add to cart
Quantity:
5 mg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,178.00
EA
Add to cart
Specifications
Chemical Name or MaterialVincristine sulfate
Name Note95%
CAS2068-78-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May damage the unborn child.