Dowex™ 1X2, Thermo Scientific Chemicals
Dowex™ 1X2, Thermo Scientific Chemicals
Dowex™ 1X2, Thermo Scientific Chemicals
Dowex™ 1X2, Thermo Scientific Chemicals
Acros Organics
Dowex™ 1X2, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number ACR202955000
Price (MYR)
766.53
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
766.53
Add to cart
Chemical Identifiers
CAS7681-82-5
IUPAC Namesodium iodide
Molecular FormulaINa
InChI KeyFVAUCKIRQBBSSJ-UHFFFAOYSA-M
SMILES[Na+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Ion exchange resin
RUO – Research Use Only