Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
CAS: 584-84-9 | C9H6N2O2 | 174.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR202502500
Price (MYR)
600.00
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
600.00
Add to cart
Chemical Identifiers
CAS14719-83-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow to cream
Melting Point (clear melt)77.5-83.5?C
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder