Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
CAS: 584-84-9 | C9H6N2O2 | 174.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Packaging:
Glass bottle
Catalog number ACR202500025
Price (MYR)
2,623.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,623.00
Add to cart
Chemical Identifiers
CAS832695-88-2
IUPAC Name[3-(methylcarbamoyl)phenyl]boronic acid
Molecular FormulaC8H10BNO3
InChI KeyFYFFPNFUVMBPRZ-UHFFFAOYSA-N
SMILESCNC(=O)C1=CC=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%