Tetrakis(triphenylphosphine)palladium(0), 99%, Thermo Scientific Chemicals
Tetrakis(triphenylphosphine)palladium(0), 99%, Thermo Scientific Chemicals
Tetrakis(triphenylphosphine)palladium(0), 99%, Thermo Scientific Chemicals
Tetrakis(triphenylphosphine)palladium(0), 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrakis(triphenylphosphine)palladium(0), 99%, Thermo Scientific Chemicals
CAS: 14221-01-3 | C72H60P4Pd | 1155.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR202380050
Price (MYR)
1,537.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,537.00
Add to cart
Chemical Identifiers
CAS9004-53-9
IUPAC Name(3R,4S,5S,6R)-2-{[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC18H32O16
InChI KeyFYGDTMLNYKFZSV-MRCIVHHJSA-N
SMILESOC[C@H]1OC(O[C@H]2[C@H](O)[C@@H](O)C(O[C@H]3[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Loss on Drying<11.0% (105°C/constant wt.)
Sulfated Ash≤1.0%