Pararosaniline base, Thermo Scientific Chemicals
Pararosaniline base, Thermo Scientific Chemicals
Pararosaniline base, Thermo Scientific Chemicals
Pararosaniline base, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pararosaniline base, Thermo Scientific Chemicals
CAS: 467-62-9 | C19H19N3O | 305.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number ACR201550100
Price (MYR)
612.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
612.00
Add to cart
Chemical Identifiers
CAS13734-34-4
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoic acid
Molecular FormulaC14H19NO4
InChI KeyZYJPUMXJBDHSIF-NSHDSACASA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)83.0-89.0?C
Optical Rotation24.4 ± 0.1? (C=1 in ethanol)
FormCrystals or powder or crystalline powder